KEY465193610 Key Organics / BIONET

ethyl 4-methyl-2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-5-carboxylate

  • MDL number MFCD03305517
  • Cas Number : 956393-95-6
  • Molecular Weight : 319.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65911

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0852_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart