KEY465193614 Key Organics / BIONET

N-{[2-(2,5-dimethyl-1H-pyrrol-1-yl)phenyl]methyl}-1-phenylmethanesulfonamide

  • MDL number MFCD03012021
  • Cas Number : 383147-69-1
  • Molecular Weight : 354.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65924

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-1016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart