KEY465193615 Key Organics / BIONET

N-[4-({[2-(2,5-dimethyl-1H-pyrrol-1-yl)phenyl]methyl}sulfamoyl)phenyl]acetamide

  • MDL number MFCD03102717
  • Cas Number : 691873-31-1
  • Molecular Weight : 397.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65925

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-1018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart