KEY465193617 Key Organics / BIONET

N-(4-ethynylphenyl)-4-methylbenzamide

  • MDL number MFCD03012854
  • Cas Number : 439095-39-3
  • Molecular Weight : 235.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-1124_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart