KEY465193624 Key Organics / BIONET

N-(4-ethynylphenyl)-2-fluorobenzene-1-sulfonamide

  • MDL number MFCD03012026
  • Cas Number : 383147-79-3
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65943

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-1149_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart