KEY465193635 Key Organics / BIONET

4-methoxy-N-[6-(morpholin-4-yl)pyridin-3-yl]benzene-1-sulfonamide

  • MDL number MFCD02580674
  • Cas Number : 439095-80-4
  • Molecular Weight : 349.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65960

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-1188_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart