KEY465193639 Key Organics / BIONET

2-[(4-chlorophenyl)sulfanyl]-5,6,7,8-tetrahydroquinoline-3-carbonitrile

  • MDL number MFCD03012871
  • Cas Number : 439095-84-8
  • Molecular Weight : 300.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65973

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-1254_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart