KEY465193641 Key Organics / BIONET

1-[4-(3-chlorothiophen-2-yl)-6-methylpyrimidin-2-yl]-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD03617997
  • Cas Number : 860788-76-7
  • Molecular Weight : 303.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65998

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-9019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart