KEY465193645 Key Organics / BIONET

1,3-dimethyl 2-methyl-2-[2-nitro-4-(trifluoromethyl)phenyl]propanedioate

  • MDL number MFCD03617898
  • Cas Number : 860788-98-3
  • Molecular Weight : 335.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66010

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart