KEY465193646 Key Organics / BIONET

4-(2-fluorophenyl)-3-methyl-1-[(4-nitrophenyl)methyl]-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD03787690
  • Cas Number : 860789-04-4
  • Molecular Weight : 328.303

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66017

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0206_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart