KEY465193647 Key Organics / BIONET

ethyl 1-acetyl-4-methyl-5-phenyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD03617900
  • Cas Number : 222403-54-5
  • Molecular Weight : 271.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66020

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0212_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart