KEY465193650 Key Organics / BIONET

4-{4-[2-(cyclohex-1-en-1-yl)ethyl]-5-methyl-4H-1,2,4-triazol-3-yl}-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD03787511
  • Cas Number : 956204-39-0
  • Molecular Weight : 286.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66032

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0228_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart