KEY465193652 Key Organics / BIONET

1,5-dimethyl (2E,4Z)-4-[(benzylamino)methylidene]pent-2-enedioate

  • MDL number MFCD03617903
  • Cas Number : 78408-80-7
  • Molecular Weight : 275.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66039

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0236_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart