KEY465193657 Key Organics / BIONET

4-{6,8-dimethoxy-4-methyl-1H,2H,3H-pyrrolo[3,2-c]quinolin-1-yl}phenol

  • MDL number MFCD03787517
  • Cas Number : 860789-35-1
  • Molecular Weight : 336.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66073

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0291_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart