KEY465193660 Key Organics / BIONET

(2E)-2-amino-3-(2,5-dimethyl-1H-pyrrol-1-yl)but-2-enedinitrile

  • MDL number MFCD03617912
  • Cas Number : 860789-50-0
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66092

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0324_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart