KEY465193662 Key Organics / BIONET

(2E)-1-(6,7-dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)-3-[(4-methoxyphenyl)amino]prop-2-en-1-one

  • MDL number MFCD03617915
  • Cas Number : 860789-54-4
  • Molecular Weight : 380.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0337_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart