KEY465193669 Key Organics / BIONET

(3E)-3-[(3-chlorophenyl)methylidene]-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD03617817
  • Cas Number : 1262616-91-0
  • Molecular Weight : 270.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66125

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0648_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart