KEY465193671 Key Organics / BIONET

(3E)-3-[(2-bromophenyl)methylidene]-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD03617818
  • Cas Number : 1262616-92-1
  • Molecular Weight : 315.166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66127

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0650_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart