KEY465193680 Key Organics / BIONET

2-[4-(4-methoxyphenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile

  • MDL number MFCD03787285
  • Cas Number : 861206-05-5
  • Molecular Weight : 244.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66174

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1W-0846_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart