KEY465193683 Key Organics / BIONET

methyl 5-bromo-2-methoxy-4-(1H-pyrrol-1-yl)benzoate

  • MDL number MFCD03617827
  • Cas Number : 861206-27-1
  • Molecular Weight : 310.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66199

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0878_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart