KEY465193685 Key Organics / BIONET

1-[1-(4-chlorobenzoyl)-1H-indol-3-yl]ethan-1-one

  • MDL number MFCD03617938
  • Cas Number : 336188-01-3
  • Molecular Weight : 297.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0899_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart