KEY465193691 Key Organics / BIONET

3-methyl-1-[(4-nitrophenyl)methyl]-4-[3-(trifluoromethyl)phenyl]-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD03787316
  • Cas Number : 861206-77-1
  • Molecular Weight : 378.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66264

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0962_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart