KEY465193692 Key Organics / BIONET

(2Z,4Z)-1,1,1,5,5,5-hexafluoro-4-[2-(3-nitropyridin-2-yl)hydrazin-1-ylidene]pent-2-en-2-ol

  • MDL number MFCD03787706
  • Cas Number : 861206-78-2
  • Molecular Weight : 344.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66265

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0963_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart