KEY465193693 Key Organics / BIONET

2-(3,5-dimethyl-4-propyl-1H-pyrazol-1-yl)-3-nitropyridine

  • MDL number MFCD03617833
  • Cas Number : 956625-18-6
  • Molecular Weight : 260.297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66267

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0965_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart