KEY465193695 Key Organics / BIONET

N,N-dimethyl-3-phenyl-5-(pyridine-3-carbonyl)thiophen-2-amine

  • MDL number MFCD27924207
  • Cas Number : 1797606-58-6
  • Molecular Weight : 308.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 122
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402878

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-7010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart