KEY465193696 Key Organics / BIONET

ethyl 1-ethyl-4-(4-fluorophenoxy)-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD11501695
  • Cas Number : 1092344-19-8
  • Molecular Weight : 343.358

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66271

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart