KEY465193698 Key Organics / BIONET

5-(thiophene-2-sulfonyl)-7-(trifluoromethyl)-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline

  • MDL number MFCD04124719
  • Cas Number : 861206-82-8
  • Molecular Weight : 388.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66273

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0203_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart