KEY465193704 Key Organics / BIONET

2-[4-(morpholin-4-yl)-3-oxobutyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD04124736
  • Cas Number : 861206-95-3
  • Molecular Weight : 302.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66292

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0230_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart