KEY465193705 Key Organics / BIONET

3-[(4E)-4-[(2H-1,3-benzodioxol-5-yl)methylidene]-5-oxo-4,5-dihydro-1H-imidazol-2-yl]-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD04124739
  • Cas Number : 1008867-63-7
  • Molecular Weight : 347.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66295

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0235_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart