KEY465193707 Key Organics / BIONET

4-(4-fluorophenyl)-5-[2-(morpholin-4-yl)ethyl]pyrimidin-2-amine

  • MDL number MFCD05668744
  • Cas Number : 861206-99-7
  • Molecular Weight : 302.353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66298

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0240_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart