KEY465193708 Key Organics / BIONET

N'-(2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carbonyl)-3-(trifluoromethyl)benzohydrazide

  • MDL number MFCD04124746
  • Cas Number : 861207-04-7
  • Molecular Weight : 393.322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66304

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0249_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart