KEY465193709 Key Organics / BIONET

5-(4-fluorophenyl)-6-[2-(morpholin-4-yl)ethyl]-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine hydrate

  • MDL number MFCD04124749
  • Cas Number : 1052545-36-4
  • Molecular Weight : 360.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66308

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0261_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart