KEY465193710 Key Organics / BIONET

5-[(4-fluorophenyl)methyl]-6-methyl-2-(5-oxo-3-phenyl-2,5-dihydro-1H-pyrazol-1-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD04124751
  • Cas Number : 861207-08-1
  • Molecular Weight : 376.391

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66310

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0265_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart