KEY465193714 Key Organics / BIONET

2-({2-[2-(3-chlorophenyl)-1,3-thiazol-4-yl]ethyl}amino)-5-nitrobenzonitrile

  • MDL number MFCD04124758
  • Cas Number : 861207-14-9
  • Molecular Weight : 384.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66318

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0281_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart