KEY465193719 Key Organics / BIONET

5-(3-fluoro-4-methoxyphenyl)-6-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD04124782
  • Cas Number : 861207-38-7
  • Molecular Weight : 273.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66342

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0311_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart