KEY465193720 Key Organics / BIONET

1-[(5-chloro-3-methyl-1-benzothiophen-2-yl)sulfonyl]-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD04124784
  • Cas Number : 861207-40-1
  • Molecular Weight : 386.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0313_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart