KEY465193721 Key Organics / BIONET

1-(2,4-dichloro-5-methylbenzenesulfonyl)-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD04124785
  • Cas Number : 861207-41-2
  • Molecular Weight : 365.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66345

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0314_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart