KEY465193734 Key Organics / BIONET

(2E)-3-(3-chloro-4-methoxyphenyl)-1-(4-nitrophenyl)prop-2-en-1-one

  • MDL number MFCD04124808
  • Cas Number : 861207-61-6
  • Molecular Weight : 317.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66388

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0834_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart