KEY465193736 Key Organics / BIONET

(3E)-4-{4-[(6-chloropyridin-3-yl)methoxy]phenyl}but-3-en-2-one

  • MDL number MFCD04124812
  • Cas Number : 861207-64-9
  • Molecular Weight : 287.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66392

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0839_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart