KEY465193742 Key Organics / BIONET

1-(1-cyclohexanecarbonyl-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD04124819
  • Cas Number : 861207-70-7
  • Molecular Weight : 269.344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66402

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0853_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart