KEY465193746 Key Organics / BIONET

2-[(E)-[(3-chloro-4-methoxyphenyl)methylidene]amino]ethan-1-ol

  • MDL number MFCD04124828
  • Cas Number : 861207-77-4
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66414

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0871_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart