KEY465193748 Key Organics / BIONET

1-[(6-chloropyridin-3-yl)methyl]-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD05668747
  • Cas Number : 861207-79-6
  • Molecular Weight : 273.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66417

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0874_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart