KEY465193749 Key Organics / BIONET

1-[(6-chloropyridin-3-yl)methyl]-4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD04124831
  • Cas Number : 159461-19-5
  • Molecular Weight : 275.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66418

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0875_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart