KEY465193750 Key Organics / BIONET

tert-butyl 2-(1,4-dimethyl-9H-carbazol-9-yl)acetate

  • MDL number MFCD04124835
  • Cas Number : 861207-83-2
  • Molecular Weight : 309.409

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66423

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0881_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart