KEY465193754 Key Organics / BIONET

[(E)-({4-[(6-chloropyridin-3-yl)methoxy]-3-methoxyphenyl}methylidene)amino]thiourea

  • MDL number MFCD04124842
  • Cas Number : 861207-89-8
  • Molecular Weight : 350.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66430

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0891_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart