KEY465193759 Key Organics / BIONET

6'-methoxy-1-methyl-2',3',4',9'-tetrahydrospiro[piperidine-4,1'-pyrido[3,4-b]indole] dihydrochloride

  • MDL number MFCD04124848
  • Cas Number : 5997-39-7
  • Molecular Weight : 358.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66436

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0899_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart