KEY465193762 Key Organics / BIONET

2-[(2-phenoxyethyl)sulfanyl]acetic acid

  • MDL number MFCD00085152
  • Cas Number : 75434-70-7
  • Molecular Weight : 212.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66448

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0927_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart