KEY465193764 Key Organics / BIONET

[3-(2,3-dihydro-1H-inden-5-yl)-1-phenyl-1H-pyrazol-4-yl]methanol

  • MDL number MFCD06496129
  • Cas Number : 956786-77-9
  • Molecular Weight : 290.366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66461

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart