KEY465193768 Key Organics / BIONET

8-(4-fluorophenyl)-5,12,14-trioxa-2-azatetracyclo[7.7.0.0^{3,7}.0^{11,15}]hexadeca-1(9),2,7,10,15-pentaen-6-one

  • MDL number MFCD06496140
  • Cas Number : 861208-11-9
  • Molecular Weight : 323.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66479

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Y-0274_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart