KEY465193772 Key Organics / BIONET

4,5-dimethyl 1-(4,6-dimethylpyrimidin-2-yl)-1H-1,2,3-triazole-4,5-dicarboxylate

  • MDL number MFCD00995003
  • Cas Number : 23951-57-7
  • Molecular Weight : 291.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66522

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Y-0824_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart